4-Hydroxy-3,5-dimethylbenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13051-14
A13051-03
A13051-06
CAAAAA13051-14EA
851.62
CAD
CAAAAA13051-14
CAAAA13051-03
CAAAA13051-06
4-Hydroxy-3,5-dimethylbenzoic acid 98%
4-Hydroxy-3,5-dimethylbenzoic acid
Purity: 98%
CAS No.: 4919-37-3
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 221-226°
Beilstein Reference: 2576289
Formula:
C₉H₁₀O₃ Melting Pt: 221…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016536 CAS Number: 4919-37-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...