L(+)-Lysine monohydrate 97%
Supplier: Thermo Scientific Chemicals
A13040-14
A13040-22
A13040-36
CAAAA13040-14EA
94.33
CAD
CAAAA13040-14
CAAAA13040-22
CAAAA13040-36
L(+)-Lysine monohydrate 97%
L(+)-Lysine monohydrate
Purity: 97%
CAS No.: 39665-12-8
Molecular Formula: C6H14N2O2.H2O
Formula Weight: 164.21 (146.19anhy)
Melting Point: 210° dec.
Beilstein Reference: 1722531
Merck Reference: 14,5636
Optical Rotation: +22.5° (c=2 in 6M HCl)
MDL No.: MFCD00151035
Formula:
H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O Melting Pt: 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00151035 CAS Number: 39665-12-8 EINECS: 200-294-2 Merck Index: 14,5636 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...