2,5-Dichlorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,4-Dichloro-2-methylbenzene
A13039-14
A13039-22
CAAAA13039-06EA
98.15
CAD
CAAAA13039-06
CAAAA13039-14
CAAAA13039-22
2,5-Dichlorotoluene 98%
2,5-Dichlorotoluene
Purity: 98%
CAS No.: 19398-61-9
Molecular Formula: C7H6Cl2
Formula Weight: 161.03
Flash Point: 79°(174°F)
Refractive Index: 1.5470
Melting Point: 4-5°
Boiling Point: 198-200°
Beilstein Reference: 1859112
UN No.: 3082
MDL No.: MFCD00000609
Formula:
C₇H₆Cl₂ MW: 161.03 g/mol Boiling Pt: 198…200 °C Melting Pt: 4…5 °C Density: 1.256 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000609 CAS Number: 19398-61-9 EINECS: 243-032-2 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...