4-Fluorobenzonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
p-Fluorobenzonitrile
A13028-06
A13028-14
A13028-22
CAAAA13028-06EA
43.29
CAD
CAAAA13028-06
CAAAA13028-14
CAAAA13028-22
4-Fluorobenzonitrile 99%
4-Fluorobenzonitrile
Purity: 99%
CAS No.: 1194-02-1
Molecular Formula: C7H4FN
Formula Weight: 121.11
Flash Point: 65°(149°F)
Odor: Almond-like
Boiling Point: 186-188°
Beilstein Reference: 2041517
MDL No.: MFCD00001812
Formula:
C₇H₄FN MW: 121.11 g/mol Boiling Pt: 186…188 °C Melting Pt: 34…37 °C Density: ~ 1 g/cm³ (20 °C) Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001812 CAS Number: 1194-02-1 EINECS: 214-784-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...