Cinnamyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cinnamic alcohol, 3-Phenylprop-2-en-1-ol
A13025-0B
A13025-30
CAAAA13025-0BEA
192.76
CAD
CAAAA13025-0B
CAAAA13025-30
Cinnamyl alcohol 98%
Cinnamyl alcohol
Purity: 98%
CAS No.: 104-54-1
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 126°(258°F)
Odor: Pleasant
Boiling Point: 257-258°
Beilstein Reference: 1903999
Merck Reference: 14,2302
MDL No.: MFCD00002921
Formula:
C₉H₁₀O MW: 134.18 g/mol Boiling Pt: 257…258 °C Melting Pt: 32…35 °C Density: 1.044 Flash Pt: 126 °C(258 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002921 CAS Number: 104-54-1 EINECS: 203-212-3 Merck Index: 14,2302 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...