Aniline hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylamine HCl, Aminobenzene hydrochloride, Anilinium chloride, Aminobenzene HCl, Phenylamine hydrochloride

A13024-0E A13024-22 A13024-36
CAAAAA13024-0EEA 609.88 CAD
CAAAAA13024-0E CAAAA13024-22 CAAAA13024-36
Aniline hydrochloride 99%
Aniline hydrochloride

Purity: 99% CAS No.: 142-04-1 Molecular Formula: C6H7N.HCl Formula Weight: 129.59 Flash Point: 193°(379°F) Melting Point: 196-200° Boiling Point: 245° Beilstein Reference: 3593823 Merck Reference: 14,659 UN No.: 1548 MDL No.: MFCD00012958

Formula: C₆H₈ClN
MW: 129.59 g/mol
Boiling Pt: 245
Melting Pt: 198
Density: 1.221
Flash Pt: 193 °C (379 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012958
CAS Number: 142-04-1
EINECS: 205-519-8
UN: 1548
ADR: 6.1 III
Merck Index: 14,659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR