3-Phenyl-1-propanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Phenyl-1-Propanol

A13022-0B A13022-0I A13022-30
CAAAA13022-0BEA 206.98 CAD
CAAAA13022-0B CAAAA13022-0I CAAAA13022-30
3-Phenyl-1-propanol 99%
3-Phenyl-1-propanol

Purity: 99% CAS No.: 122-97-4 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 109°(228°F) Refractive Index: 1.5260 Melting Point: -18° Boiling Point: 235° Beilstein Reference: 1857542 MDL No.: MFCD00002950

Formula: (C₆H₅)CH₂CH₂CH₂OH
Boiling Pt: 235 °C
Melting Pt: –18 °C
Density: 1.004
Flash Pt: 109 °C(228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002950
CAS Number: 122-97-4
EINECS: 204-587-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR