2-Aminoisobutyric acid 99%

Supplier: Thermo Scientific Chemicals

A13021-14 A13021-22 A13021-30
CAAAAA13021-14EA 64.94 CAD
CAAAAA13021-14 CAAAAA13021-22 CAAAA13021-30
2-Aminoisobutyric acid 99%
2-Aminoisobutyric acid
Formula: C₄H₉NO₂
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: >300 °C
Density: 1.278 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008049
CAS Number: 62-57-7
EINECS: 200-544-0
Merck Index: 14,445

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR