2,6-Difluoroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13020-06
A13020-14
A13020-22
CAAAA13020-06EA
68.73
CAD
CAAAA13020-06
CAAAA13020-14
CAAAA13020-22
2,6-Difluoroaniline 98%
2,6-Difluoroaniline
Purity: 98%
CAS No.: 5509-65-9
Molecular Formula: C6H5F2N
Formula Weight: 129.11
Flash Point: 51°(124°F)
Refractive Index: 1.5080
Odor: Characteristic
Boiling Point: 152-154°
Beilstein Reference: 2802697
UN No.: 1992
MDL No.: MFCD00007655
Formula:
C₆H₅F₂N Boiling Pt: 152…154 °C Density: 1.275 Flash Pt: 51 °C (124 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007655 CAS Number: 5509-65-9 EINECS: 226-853-0 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...