4-(Dimethylamino)pyridine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine

A13016-06 A13016-14 A13016-22
CAAAA13016-06EA 38.55 CAD
CAAAA13016-06 CAAAA13016-14 CAAAA13016-22
4-(Dimethylamino)pyridine 99%
4-(Dimethylamino)pyridine

Purity: 99% CAS No.: 1122-58-3 Molecular Formula: C7H10N2 Formula Weight: 122.17 Flash Point: 124°(255°F) Physical Form: Crystals or powder Melting Point: 110-113° Boiling Point: 162°/50mm Beilstein Reference: 110354 UN No.: 2928 MDL No.: MFCD00006418

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 195
Melting Pt: 110…113 °C
Flash Pt: 124 °C (255 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006418
CAS Number: 1122-58-3
EINECS: 214-353-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR