p-Anisohydrazide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13015-09 A13015-18 A13015-30
CAAAA13015-09EA 57.04 CAD
CAAAA13015-09 CAAAA13015-18 CAAAA13015-30
p-Anisohydrazide 98+%
p-Anisohydrazide

Purity: 98+% CAS No.: 3290-99-1 Molecular Formula: C8H10N2O2 Formula Weight: 166.18 Melting Point: 136-140° Beilstein Reference: 389017

Formula: C₈H₁₀N₂O₂
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00017073
CAS Number: 3290-99-1
EINECS: 221-952-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR