1-Bromo-4-ethylbenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Bromophenyl)ethane
A13013-09
A13013-18
CAAAAA13013-30EA
1366.19
CAD
CAAAAA13013-30
CAAAA13013-09
CAAAA13013-18
1-Bromo-4-ethylbenzene 99%
1-Bromo-4-ethylbenzene
Purity: 99%
CAS No.: 1585-07-5
Molecular Formula: C8H9Br
Formula Weight: 185.06
Flash Point: 63°(145°F)
Refractive Index: 1.5440
Boiling Point: 203-204°
Beilstein Reference: 1856989
MDL No.: MFCD00000111
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 203…204 °C Melting Pt: –43,5 °C Density: 1,341 Flash Pt: 63 °C(145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000111 CAS Number: 1585-07-5 EINECS: 216-439-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...