5-Nitrosalicylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13010-06
A13010-14
CAAAA13010-06EA
43.13
CAD
CAAAA13010-06
CAAAA13010-14
5-Nitrosalicylic acid 98%
5-Nitrosalicylic acid
Formula:
C₇H₅NO₅ Melting Pt: 228…236 °C Density: 1.650 Storage Temperature: Ambient |
MDL Number:
MFCD00007338 CAS Number: 96-97-9 EINECS: 202-548-8 Merck Index: 14,6632 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...