2-Pentyn-1-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12999-03
A12999-06
A12999-14
CAAAA12999-03EA
35.55
CAD
CAAAA12999-03
CAAAA12999-06
CAAAA12999-14
2-Pentyn-1-ol 98%
2-Pentyn-1-ol
Purity: 98%
CAS No.: 6261-22-9
Molecular Formula: C5H8O
Formula Weight: 84.12
Storage Temperature: 2°C to 8°C
Flash Point: 58°(136°F)
Refractive Index: 1.4520
Odor: Recognizable
Boiling Point: 154°
Beilstein Reference: 1734007
UN No.: 1987
MDL No.: MFCD00040915
Formula:
C₅H₈O Boiling Pt: 154 °C Melting Pt: –0.2 °C Density: 0.910 Flash Pt: 58 °C (136 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00040915 CAS Number: 6261-22-9 EINECS: 228-411-2 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...