2,3-Dibromothiophene 98+%

Supplier: Thermo Scientific Chemicals
Warning

A12998-06 A12998-14
CAAAA12998-03EA 36.91 CAD
CAAAA12998-03 CAAAA12998-06 CAAAA12998-14
2,3-Dibromothiophene 98+%
2,3-Dibromothiophene

Purity: 98+% CAS No.: 3140-93-0 Molecular Formula: C4H2Br2S Formula Weight: 241.94 Flash Point: 100°(212°F) Refractive Index: 1.6310 Boiling Point: 215-216° Beilstein Reference: 107512 MDL No.: MFCD00005418

Formula: C₄H₂Br₂S
Boiling Pt: 215…216 °C
Melting Pt: –17.5 °C
Density: 2.135
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005418
CAS Number: 3140-93-0
EINECS: 221-542-6
UN: 1992
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR