Anisole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl phenyl ether, Methoxybenzene
A12997-0E
A12997-36
CAAAA12997-0EEA
238.58
CAD
CAAAA12997-0E
CAAAA12997-36
Anisole 99%
Anisole
Purity: 99%
CAS No.: 100-66-3
Molecular Formula: C7H8O
Formula Weight: 108.14
Flash Point: 43°(109°F)
Refractive Index: 1.5170
Odor: Ether-like
Melting Point: -37°
Boiling Point: 153-154°
Beilstein Reference: 506892
Merck Reference: 14,669
UN No.: 2222
MDL No.: MFCD00008354
Formula:
C₇H₈O MW: 108.14 g/mol Boiling Pt: 154 Melting Pt: -37 Density: 0.994 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008354 CAS Number: 100-66-3 EINECS: 202-876-1 UN: 2222 ADR: 3 III Merck Index: 14,669 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...