2',4'-Dimethylacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(2,4-Dimethylphenyl)ethan-1-one, 2,4-Dimethylacetophenone, 2,4-Dimethylacetophenon
A12992-09
A12992-18
A12992-30
CAAAA12992-09EA
59.72
CAD
CAAAA12992-09
CAAAA12992-18
CAAAA12992-30
2',4'-Dimethylacetophenone 97%
2',4'-Dimethylacetophenone
Purity: 97%
CAS No.: 89-74-7
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 100°(212°F)
Refractive Index: 1.5350
Odor: Characteristic
Boiling Point: 227-228°
Beilstein Reference: 956772
MDL No.: MFCD00003571
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 227…228 °C Density: 0.999 Flash Pt: 100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003571 CAS Number: 89-74-7 EINECS: 201-935-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...