4'-Nitrocinnamic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A12990-14 A12990-22
CAAAAA12990-36EA 947.98 CAD
CAAAAA12990-36 CAAAA12990-14 CAAAA12990-22
4'-Nitrocinnamic acid 98+%
4'-Nitrocinnamic acid

Purity: 98+% CAS No.: 882-06-4 Molecular Formula: C9H7NO4 Formula Weight: 193.16 Odor: Odorless Melting Point: ca 290° dec. Beilstein Reference: 1369659 MDL No.: MFCD00007381

Formula: C₉H₇NO₄
Melting Pt: ∼ 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007381
CAS Number: 1866-39-3
EINECS: 212-924-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR