(±)-4-Bromomandelic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12978-06
A12978-14
A12978-22
CAAAA12978-06EA
82.48
CAD
CAAAA12978-06
CAAAA12978-14
CAAAA12978-22
(±)-4-Bromomandelic acid 98+%
(±)-4-Bromomandelic acid
Purity: 98+%
CAS No.: 6940-50-7
Molecular Formula: C8H7BrO3
Formula Weight: 231.05
Melting Point: 115-119°
Beilstein Reference: 1567909
Merck Reference: 14,1424
MDL No.: MFCD00004232
Formula:
C₈H₇BrO₃ Boiling Pt: 344 °C (1013 hPa) Melting Pt: 115…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004232 CAS Number: 6940-50-7 EINECS: 230-085-1 Merck Index: 14,1424 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...