(1-Propyl)triphenylphosphonium bromide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12968-0B
A12968-18
A12968-30
CAAAAA12968-0BEA
978.02
CAD
CAAAAA12968-0B
CAAAA12968-18
CAAAA12968-30
(1-Propyl)triphenylphosphonium bromide 99%
(1-Propyl)triphenylphosphonium bromide
Purity: 99%
CAS No.: 6228-47-3
Molecular Formula: CH3(CH2)2P(C6H5)3Br
Formula Weight: 385.29
Melting Point: 236-240°
Beilstein Reference: 3599726
Formula:
C₂₁H₂₂BrP Melting Pt: 236…240 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011843 CAS Number: 6228-47-3 EINECS: 228-330-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...