Allylbenzene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12965-14
A12965-22
CAAAA12965-14EA
116.29
CAD
CAAAA12965-14
CAAAA12965-22
Allylbenzene 98%
Allylbenzene
Purity: 98%
CAS No.: 300-57-2
Molecular Formula: C9H10
Formula Weight: 118.18
Flash Point: 33°(91°F)
Refractive Index: 1.5100
Odor: Aromatic
Melting Point: ca -50°
Boiling Point: 158-160°
Beilstein Reference: 1098501
UN No.: 3295
MDL No.: MFCD00008651
Formula:
(C₆H₅)CH₂CHCH₂ Boiling Pt: 158…160 °C Melting Pt: ∼ 50 °C Density: 0.892 Flash Pt: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008651 CAS Number: 300-57-2 EINECS: 206-095-7 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...