2-Bromo-1,4-difluorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,5-Difluorobromobenzene, 1-Bromo-2,5-difluorobenzene

A12963-06 A12963-14 A12963-22
CAAAA12963-06EA 106.18 CAD
CAAAA12963-06 CAAAA12963-14 CAAAA12963-22
2-Bromo-1,4-difluorobenzene 98%
2-Bromo-1,4-difluorobenzene

Purity: 98% CAS No.: 399-94-0 Molecular Formula: C6H3BrF2 Formula Weight: 192.99 Flash Point: 65°(149°F) Refractive Index: 1.5080 Melting Point: -31° Boiling Point: 73-75°/65mm Beilstein Reference: 1680893 MDL No.: MFCD00000345

Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 73…75 °C/65 mm
Melting Pt: –31 °C
Density: 1.714
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000345
CAS Number: 399-94-0
EINECS: 206-920-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR