2-Propylvaleric acid 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12962-09
A12962-18
A12962-30
CAAAA12962-09EA
62.57
CAD
CAAAA12962-09
CAAAA12962-18
CAAAA12962-30
2-Propylvaleric acid 98+%
2-Propylvaleric acid
Purity: 98+%
CAS No.: 99-66-1
Molecular Formula: C8H16O2
Formula Weight: 144.21
Flash Point: 126°(258°F)
Refractive Index: 1.4250
Odor: Weak, characteristic
Boiling Point: 219-220°
Beilstein Reference: 1750447
Merck Reference: 14,9913
UN No.: 3265
MDL No.: MFCD00002672
Formula:
C₈H₁₆O₂ Boiling Pt: 219…220 °C Density: 0.905 Flash Pt: 126 °C (258 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002672 CAS Number: 99-66-1 EINECS: 202-777-3 UN: 3265 ADR: 8,III Merck Index: 14,9913 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...