Adamantane-1-carboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Adamantanecarboxylic acid, Tricyclo[3.3.1.1(3,7)]decane-1-carboxylic acid

A12959-36 A12959-14 A12959-22
CAAAAA12959-36EA 1251.36 CAD
CAAAAA12959-36 CAAAA12959-14 CAAAA12959-22
Adamantane-1-carboxylic acid 99%
Adamantane-1-carboxylic acid

Purity: 99% CAS No.: 828-51-3 Molecular Formula: C11H16O2 Formula Weight: 180.25 Melting Point: 173-176° Beilstein Reference: 1910637

Formula: C₁₁H₁₆O₂
MW: 180.25 g/mol
Melting Pt: 173…176 °C
Density: 1.221 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074720
CAS Number: 828-51-3
EINECS: 212-584-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR