Basic fuchsin
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
Biological stain for distinguishing coli and aerogene bacteria
CAS No.: 632-99-5
Molecular Formula: C20H20ClN3
Formula Weight: 337.86
Melting Point: 268-270° dec.
Solubility: Soluble in water
Beilstein Reference: 4166684
Merck Reference: 14,5652
MDL No.: MFCD00012569
Formula:
C₂₀H₂₀ClN₃ Melting Pt: 268…270 °C (dec.) Density: 1.22 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012569 CAS Number: 632-99-5 EINECS: 211-189-6 Merck Index: 14,5652 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...