4-Bromo-2-nitrotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning

A12951-22 A12951-06
CAAAAA12951-22EA 698.36 CAD
CAAAAA12951-22 CAAAA12951-06 CAAAA12951-14
4-Bromo-2-nitrotoluene 99%
4-Bromo-2-nitrotoluene

Purity: 99% CAS No.: 60956-26-5 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Flash Point: 110°(230°F) Melting Point: 45-48° Boiling Point: 130°/12mm Beilstein Reference: 2328190 MDL No.: MFCD00041243

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 130 °C/12 mm
Melting Pt: 45…48 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041243
CAS Number: 60956-26-5
EINECS: 262-536-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR