Tritylium tetrafluoroborate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12949-09
A12949-18
A12949-30
CAAAA12949-09EA
103.17
CAD
CAAAA12949-09
CAAAA12949-18
CAAAA12949-30
Tritylium tetrafluoroborate 98%
Tritylium tetrafluoroborate
Purity: 98%
CAS No.: 341-02-6
Molecular Formula: C19H15BF4
Formula Weight: 330.13
Storage Temperature: 2°C to 8°C
Odor: Odorless
Beilstein Reference: 5085649
UN No.: 3261
MDL No.: MFCD00013120
Formula:
C₁₉H₁₅BF₄ Melting Pt: ca 207 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013120 CAS Number: 341-02-6 EINECS: 206-433-3 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...