6-Amino-3-bromo-2-methylpyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Amino-3-bromo-2-picoline, 6-AMINO-3-BROMO-2-METHYLPYRIDINE, 3-Bromo-2-methyl-6-pyridylamine, 5-Bromo-6-methylpyridin-2-amine, 3-Bromo-2-methylpyridin-6-amine, 2-Amino-5-bromo-6-picoline, ['6-Amino-3-bromo-2-picoline'], 5-Bromo-6-methyl-2-pyridylamine, 2-Amino-5-bromo-6-methylpyridine
A12948-06
A12948-14
CAAAA12948-03EA
69
CAD
CAAAA12948-03
CAAAA12948-06
CAAAA12948-14
6-Amino-3-bromo-2-methylpyridine 97%
6-Amino-3-bromo-2-methylpyridine
Purity: 97%
CAS No.: 42753-71-9
Molecular Formula: C6H7BrN2
Formula Weight: 187.04
Melting Point: 80-82°
Beilstein Reference: 114140
Formula:
C₆H₇BrN₂ MW: 187.04 g/mol Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068230 CAS Number: 42753-71-9 EINECS: 255-927-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...