4-Fluorobenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
p-Fluorobenzoyl chloride
A12946-14
A12946-22
A12946-36
CAAAA12946-14EA
77.89
CAD
CAAAA12946-14
CAAAA12946-22
CAAAA12946-36
4-Fluorobenzoyl chloride 98%
4-Fluorobenzoyl chloride
Purity: 98%
CAS No.: 403-43-0
Molecular Formula: C7H4ClFO
Formula Weight: 158.56
Flash Point: 82°(179°F)
Refractive Index: 1.5320
Odor: Acidic
Melting Point: 10-12°
Boiling Point: 193°
Beilstein Reference: 386215
UN No.: 3265
MDL No.: MFCD00000684
Formula:
C₇H₄ClFO MW: 158.56 g/mol Boiling Pt: 180 Melting Pt: 9 Density: 1.326 Flash Pt: 82 °C (179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000684 CAS Number: 403-43-0 EINECS: 206-961-4 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...