2-Bromopropane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12944-AK
A12944-AU
A12944-0F
CAAAA12944-0JEA
1916.65
CAD
CAAAA12944-0J
CAAAA12944-AK
CAAAA12944-AU
AAA12944-0F
AAA12944-L8
2-Bromopropane 99%
2-Bromopropane
Purity: 99%
CAS No.: 75-26-3
Molecular Formula: C3H7Br
Formula Weight: 123.00
Flash Point: 19°(66°F)
Physical Form: Liquid
Density: 1.310
Refractive Index: 1.4255
Appearance: Colorless to pale yellow
Melting Point: -89°
Boiling Point: 59-60°
Beilstein Reference: 741852
Merck Reference: 14,5210
UN No.: 2344
MDL No.: MFCD00000147
Formula:
H₃CBrCHCH₃ Boiling Pt: 59…60 °C Melting Pt: –89 °C Density: 1.310 Flash Pt: 19 °C(66 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000147 CAS Number: 75-26-3 EINECS: 200-855-1 UN: 2344 ADR: "3",III Merck Index: 14,5210 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...