1-Methylisatin 97%

Supplier: Thermo Scientific Chemicals
Warning

A12942-03 A12942-06
CAAAA12942-03EA 63.2 CAD
CAAAA12942-03 CAAAA12942-06 CAAAA12942-14
1-Methylisatin 97%
1-Methylisatin

Purity: 97% CAS No.: 2058-74-4 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: 129-133° Beilstein Reference: 128280

Formula: C₉H₇NO₂
Melting Pt: 129…133 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005812
CAS Number: 2058-74-4
EINECS: 218-164-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR