(4-Hydroxyphenoxy)acetic acid 98+%
Supplier: Thermo Scientific Chemicals
A12940-06
A12940-14
A12940-22
CAAAA12940-06EA
109.65
CAD
CAAAA12940-06
CAAAA12940-14
CAAAA12940-22
(4-Hydroxyphenoxy)acetic acid 98+%
(4-Hydroxyphenoxy)acetic acid
Purity: 98+%
CAS No.: 1878-84-8
Molecular Formula: C8H8O4
Formula Weight: 168.15
Melting Point: 153-158°
Beilstein Reference: 1952779
Formula:
(C₆H₄(OH))OCH₂COOH |
MDL Number:
MFCD00014362 CAS Number: 1878-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...