Methyl-4-nitrobenzoate 99%

Supplier: Thermo Scientific Chemicals

A12928-22 A12928-36
CAAAAA12928-0EEA 1143.16 CAD
CAAAAA12928-0E CAAAA12928-22 CAAAA12928-36
Methyl-4-nitrobenzoate 99%
Methyl 4-nitrobenzoate

Purity: 99% CAS No.: 619-50-1 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 95-97° Beilstein Reference: 1912198

Formula: C₈H₇NO₄
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00007350
CAS Number: 619-50-1
EINECS: 210-599-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR