4-Chloro-1,2-difluorobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,4-Difluorochlorobenzene
A12924-06
A12924-14
A12924-22
CAAAA12924-06EA
66.36
CAD
CAAAA12924-06
CAAAA12924-14
CAAAA12924-22
4-Chloro-1,2-difluorobenzene 98%
4-Chloro-1,2-difluorobenzene
Purity: 98%
CAS No.: 696-02-6
Molecular Formula: C6H3ClF2
Formula Weight: 148.54
Flash Point: 32°(89°F)
Refractive Index: 1.4745
Odor: Aromatic
Boiling Point: 128-130°
Beilstein Reference: 2081081
UN No.: 1993
MDL No.: MFCD00042572
Formula:
C₆H₃ClF₂ MW: 148.54 g/mol Boiling Pt: 128…130 °C Density: 1.350 Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042572 CAS Number: 696-02-6 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...