3,5-Dichloro-4-hydroxybenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

A12923-06 A12923-14 A12923-22
CAAAA12923-06EA 51.35 CAD
CAAAA12923-06 CAAAA12923-14 CAAAA12923-22
3,5-Dichloro-4-hydroxybenzoic acid 97%
3,5-Dichloro-4-hydroxybenzoic acid

Purity: 97% CAS No.: 3336-41-2 Molecular Formula: C7H4Cl2O3 Formula Weight: 207.01 Melting Point: 265-270° Beilstein Reference: 2616297

Formula: C₇H₄Cl₂O₃
Boiling Pt: 341 °C (1013 hPa)
Melting Pt: 265…270 °C
Storage Temperature: Ambient
MDL Number: MFCD00002550
CAS Number: 3336-41-2
EINECS: 222-071-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR