Sodium-L(+)-glutamate monohydrate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-(+)-2-Aminopentanedioic acid sodium salt monohydrate, Sodium L-Glutamate Monohydrate, L(+)-Glutamic acid sodium salt monohydrate, L(+)-2-Aminopentanedioic acid sodium salt monohydrate, (S)-(+)-Glutamic acid sodium salt monohydrate, L(+)-Glutamic acid monosodium salt monohydrate, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-Sodium glutamate monohydrate, L(+)-α-Glutamic acid sodium salt monohydrate, L(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-α-Glutamic acid sodium salt monohydrate

A12919-0B A12919-0I A12919-30
CAAAA12919-0BEA 94.82 CAD
CAAAA12919-0B CAAAA12919-0I CAAAA12919-30
Sodium-L(+)-glutamate monohydrate 98+%
Sodium-L(+)-glutamate monohydrate

Purity: 98+% CAS No.: 6106-04-3 Molecular Formula: C5H8NNaO4.H2O Formula Weight: 187.14 (169.12anhy) Melting Point: 228° dec. Beilstein Reference: 4164348

Formula: C₅H₈NNaO₄·H₂O
Melting Pt: 228 °C (dec.)
Flash Pt: ≥ 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00150138
CAS Number: 6106-04-3
EINECS: 205-538-1
Merck Index: 13,06278

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR