1-Acetylnaphthalene 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1'-Acetonaphthone, α-acetonaphthone, 1'-Acetylnaphthalene, Methyl α-naphthyl ketone, Methyl 1-naphthyl ketone, 1-Acetonaphthone

A12918-0B A12918-18 A12918-30
CAAAA12918-0BEA 597.24 CAD
CAAAA12918-0B CAAAA12918-18 CAAAA12918-30
1-Acetylnaphthalene 97+%
1-Acetylnaphthalene

Purity: 97+% CAS No.: 941-98-0 Molecular Formula: C12H10O Formula Weight: 170.21 Flash Point: 110°(230°F) Refractive Index: 1.6280 Melting Point: 9-11° Boiling Point: 302° Beilstein Reference: 1100618 MDL No.: MFCD00004013

Formula: C₁₂H₁₀O
Boiling Pt: 302 °C
Melting Pt: 9…11 °C
Density: 1.120
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004013
CAS Number: 941-98-0
EINECS: 213-384-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR