γ-(N-Morpholino)propanesulfonic acid (MOPS) 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Morpholinopropane-1-sulfonic acid, 3-(N-Morpholino)propanesulfonic acid, 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
A12914-14
A12914-22
A12914-36
CAAAA12914-14EA
62.73
CAD
CAAAA12914-14
CAAAA12914-22
CAAAA12914-36
γ-(N-Morpholino)propanesulfonic acid (MOPS) 99%
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Good's buffer
Purity: 99%
CAS No.: 1132-61-2
Molecular Formula: C7H15NO4S
Formula Weight: 209.27
Flash Point: 110°(230°F)
Physical Form: Powder
Melting Point: 277-280° dec.
Beilstein Reference: 1106776
Merck Reference: 14,6265
MDL No.: MFCD00006183
Good's buffer
Formula:
C₇H₁₅NO₄S MW: 209.27 g/mol Melting Pt: 277…280 °C (dec.) Flash Pt: 110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006183 CAS Number: 1132-61-2 EINECS: 214-478-5 Merck Index: 14,6265 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...