9-Fluorenone 98+%
Supplier: Thermo Scientific Chemicals
A12910-0E
A12910-22
A12910-36
CAAAA12910-0EEA
1181.84
CAD
CAAAA12910-0E
CAAAA12910-22
CAAAA12910-36
9-Fluorenone 98+%
9-Fluorenone
Purity: 98+%
CAS No.: 486-25-9
Molecular Formula: C13H8O
Formula Weight: 180.21
Flash Point: 163°(325°F)
Physical Form: Cryst powder or flakes
Density: 0.9
Appearance: Yellow
Melting Point: 82-85°
Boiling Point: 342°
Beilstein Reference: 1636531
MDL No.: MFCD00001141
Formula:
C₁₃H₈O Boiling Pt: 342 °C Melting Pt: 82…85 °C Density: 0.9 Flash Pt: 163 °C(325 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001141 CAS Number: 486-25-9 EINECS: 207-630-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...