Ethyl cinnamate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12906-0E
A12906-22
A12906-36
CAAAA12906-0EEA
723.4
CAD
CAAAA12906-0E
CAAAA12906-22
CAAAA12906-36
Ethyl cinnamate 98+%
Purity: 98+%
CAS No.: 103-36-6
Molecular Formula: C11H12O2
Formula Weight: 176.22
Flash Point: 135°(275°F)
Physical Form: Liquid
Density: 1.049
Refractive Index: 1.5590
Appearance: Colorless
Melting Point: 7°
Boiling Point: 270-272°
Beilstein Reference: 1238804
Merck Reference: 14,2299
MDL No.: MFCD00009189
Formula:
C₁₁H₁₂O₂ Boiling Pt: 270…272 °C Melting Pt: 7 °C Density: 1.049 Flash Pt: 135 °C(275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009189 CAS Number: 103-36-6 EINECS: 203-104-6 Merck Index: 14,2299 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...