Iodotrimethylsilane 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with copper
A12902-06
A12902-14
A12902-22
A12902-30
CAAAAA12902-06EA
72.52
CAD
CAAAAA12902-06
CAAAAA12902-14
CAAAAA12902-22
CAAAA12902-30
Iodotrimethylsilane 97% stabilized
Iodotrimethylsilane
Purity: 97%
CAS No.: 16029-98-4
Molecular Formula: C3H9ISi
Formula Weight: 200.09
Storage Temperature: -30°C to -10°C
Flash Point: -2°(28°F)
Refractive Index: 1.4760
Boiling Point: 105-106°
Beilstein Reference: 1731136
UN No.: 2924
MDL No.: MFCD00001028
Formula:
C₃H₉ISi Boiling Pt: 105…106 °C Melting Pt: < 0 °C Density: 1.440 Flash Pt: –2 °C (28 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00001028 CAS Number: 16029-98-4 EINECS: 240-171-0 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...