2,5-Dimethoxybenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A12901-06 A12901-14 A12901-22
CAAAA12901-06EA 85.64 CAD
CAAAA12901-06 CAAAA12901-14 CAAAA12901-22
2,5-Dimethoxybenzoic acid 98+%
2,5-Dimethoxybenzoic acid

Purity: 98+% CAS No.: 2785-98-0 Molecular Formula: C9H10O4 Formula Weight: 182.18 Odor: Odorless Melting Point: 75-79° Beilstein Reference: 2097993 MDL No.: MFCD00002436

Formula: C₉H₁₀O₄
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00002436
CAS Number: 2785-98-0
EINECS: 220-503-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR