Pyridine-2-carbonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Cyanopyridine, 2-Pyridinecarbonitrile, Picolinonitrile

A12900-22 A12900-36
CAAAA12900-22EA 58.78 CAD
CAAAA12900-22 CAAAA12900-36
Pyridine-2-carbonitrile 99%
Pyridine-2-carbonitrile

Purity: 99% CAS No.: 100-70-9 Molecular Formula: C6H4N2 Formula Weight: 104.11 Flash Point: 89°(192°F) Refractive Index: 1.5290 Odor: Characteristic pH: 8.4 Melting Point: 26-28° Boiling Point: 212-214° Beilstein Reference: 107710 MDL No.: MFCD00006218

Formula: C₆H₄N₂
MW: 104.11 g/mol
Boiling Pt: 212…214 °C
Melting Pt: 26…28 °C
Density: 1.081
Flash Pt: 89 °C (192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006218
CAS Number: 100-70-9
EINECS: 202-880-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR