2-Iodothiophene 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with copper
A12898-14
A12898-22
CAAAA12898-14EA
83.58
CAD
CAAAA12898-14
CAAAA12898-22
2-Iodothiophene 98% stabilized
2-Iodothiophene
Purity: 98%
CAS No.: 3437-95-4
Molecular Formula: C4H3IS
Formula Weight: 210.04
Flash Point: 71°(159°F)
Refractive Index: 1.6520
Melting Point: -40°
Boiling Point: 182°
Beilstein Reference: 104666
MDL No.: MFCD00005424
Formula:
C₄H₃IS Boiling Pt: 182 °C Melting Pt: –40 °C Density: 2.064 Flash Pt: 71 °C(159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005424 CAS Number: 3437-95-4 EINECS: 222-342-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...