3-Bromopyridine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12894-14
A12894-22
A12894-36
CAAAA12894-14EA
90.69
CAD
CAAAA12894-14
CAAAA12894-22
CAAAA12894-36
3-Bromopyridine 98+%
3-Bromopyridine
Purity: 98+%
CAS No.: 626-55-1
Molecular Formula: C5H4BrN
Formula Weight: 158.00
Flash Point: 83°(181°F)
Refractive Index: 1.5700
Odor: Characteristic
Melting Point: -27°
Boiling Point: 172-174°
Beilstein Reference: 105880
UN No.: 2810
MDL No.: MFCD00006373
Formula:
C₅H₄BrN MW: 158 g/mol Boiling Pt: 172…174 °C Melting Pt: –27 °C Density: 1.617 Flash Pt: 72 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006373 CAS Number: 626-55-1 EINECS: 210-952-0 UN: 2929 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...