Ethyl-4-(dimethylamino)benzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

A12888-14 A12888-22 A12888-36
CAAAAA12888-14EA 39.82 CAD
CAAAAA12888-14 CAAAA12888-22 CAAAA12888-36
Ethyl-4-(dimethylamino)benzoate 99%
Ethyl 4-(dimethylamino)benzoate

Purity: 99% CAS No.: 10287-53-3 Molecular Formula: C11H15NO2 Formula Weight: 193.25 Melting Point: 63-65° Boiling Point: 190-191°/14mm MDL No.: MFCD00009115

Formula: C₁₁H₁₅NO₂
Boiling Pt: 190…191 °C/14 mm
Melting Pt: 63…65 °C
Density: 1.060
Storage Temperature: Ambient
MDL Number: MFCD00009115
CAS Number: 10287-53-3
EINECS: 233-634-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR