Benzhydryl alcohol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diphenylcarbinol, Benzhydrol, Diphenylmethanol
A12884-0B
A12884-18
A12884-30
CAAAA12884-0BEA
312.84
CAD
CAAAA12884-0B
CAAAA12884-18
CAAAA12884-30
Benzhydryl alcohol 99%
Benzhydryl alcohol
Purity: 99%
CAS No.: 91-01-0
Molecular Formula: C13H12O
Formula Weight: 184.24
Flash Point: 160°(320°F)
Odor: Odorless
Boiling Point: 297-298°
Beilstein Reference: 1424379
Merck Reference: 14,1090
MDL No.: MFCD00004488
Formula:
C₁₃H₁₂O Boiling Pt: 297…298 °C Melting Pt: 64…68 °C Density: 1.054 g/cm³ (80 °C) Flash Pt: 160 °C(320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004488 CAS Number: 91-01-0 EINECS: 202-033-8 Merck Index: 14,1090 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...