4-Ethylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12883-18
A12883-30
CAAAA12883-18EA
96.85
CAD
CAAAA12883-18
CAAAA12883-30
4-Ethylpyridine 98%
4-Ethylpyridine
Purity: 98%
CAS No.: 536-75-4
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 52°(126°F)
Refractive Index: 1.5010
Odor: Unpleasant
pH: 10
Melting Point: -91°
Boiling Point: 167-168°
Beilstein Reference: 106481
Merck Reference: 14,3849
UN No.: 1993
MDL No.: MFCD00006450
Formula:
C₇H₉N MW: 107.16 g/mol Boiling Pt: 167…168 °C Melting Pt: –91 °C Density: 0.942 Flash Pt: 52 °C (126 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006450 CAS Number: 536-75-4 EINECS: 208-646-7 UN: 1992 ADR: 3,III Merck Index: 14,3849 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...