4-Chloroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12879-0B
A12879-0I
A12879-30
CAAAA12879-0BEA
188.02
CAD
CAAAA12879-0B
CAAAA12879-0I
CAAAA12879-30
4-Chloroaniline 98%
4-Chloroaniline
Purity: 98%
CAS No.: 106-47-8
Molecular Formula: C6H6ClN
Formula Weight: 127.57
Flash Point: 120°(248°F)
Melting Point: 67-72°
Boiling Point: 232°
Beilstein Reference: 471359
Merck Reference: 14,2118
UN No.: 2018
MDL No.: MFCD00007835
Formula:
C₆H₆ClN Boiling Pt: 232 °C Melting Pt: 67…72 °C Density: 1.430 Flash Pt: 120 °C (248 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007835 CAS Number: 106-47-8 EINECS: 203-401-0 UN: 2018 ADR: 6.1,II Merck Index: 14,2118 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...