2-Methylcyclohexanol cis- and trans-mixture 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12877-0F
A12877-AP
CAAAA12877-0FEA
652.54
CAD
CAAAA12877-0F
CAAAA12877-AP
2-Methylcyclohexanol cis- and trans-mixture 97%
2-Methylcyclohexanol
Purity: 97%
CAS No.: 583-59-5
Molecular Formula: C7H14O
Formula Weight: 114.19
Flash Point: 58°(136°F)
Refractive Index: 1.4620
Melting Point: -21°
Boiling Point: 163-166°
Beilstein Reference: 1847535
UN No.: 2617
MDL No.: MFCD00001443
Formula:
C₇H₁₄O Boiling Pt: 163…166 °C Melting Pt: –21 °C Density: 0.929 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001443 CAS Number: 583-59-5 EINECS: 209-512-0 UN: 2617 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...